Learn More
BIOLIFE SOLUTIONS LLC CryoStor CS10 100ml
SDP
Catalog No. NC9930384
Encompass
CryoStor CS10 a cell-specific, optimized freeze media, is designed to prepare and preserve cells in ultra low temperature environments (-70 deg.C to -196 deg.C); CryoStorCS10 is pre-formulated with 10% DMSO.
Catalog No. NC9930384 Supplier BIOLIFE SOLUTIONS LLC Supplier No. 210102
CryoStor CS10 a cell-specific, optimized freeze media, is designed to prepare and preserve cells in ultra low temperature environments (-70 deg.C to -196 deg.C); CryoStorCS10 is pre-formulated with 10% DMSO.