Learn More
Charles River Labs Cell Solutions CryoStor CS10 Biopreservation Media, 100mL Bag
Catalog No. 502256134
Encompass
CryoStor is a uniquely formulated serum-free and animal component-free cryopreservation medium containing dimethyl sulfoxide (DMSO). CryoStor is designed to preserve and protect cells during the freezing and thawing processes as well as during storage at low temperature environments (-80°C to -196°C). CryoStor is cGMP manufactured with USP grade components.
This item is not returnable. View return policy
Catalog No. 50-225-6134 Supplier Charles River Labs Cell Solutions Supplier No. BPM 210202
CryoStor is a uniquely formulated serum-free and animal component-free cryopreservation medium containing dimethyl sulfoxide (DMSO). CryoStor is designed to preserve and protect cells during the freezing and thawing processes as well as during storage at low temperature environments (-80°C to -196°C). CryoStor is cGMP manufactured with USP grade components.