Learn More
BIOLIFE SOLUTIONS LLC CryoStor CS10, 100 mL, pre-formulated, serum-free, protein-free, animal component-free, 10% DMSO, ready-to-use
SDP
Catalog No. NC3074110
Encompass
CryoStor CS10, 100 mL, pre-formulated, serum-free, protein-free, animal component-free, 10% DMSO, ready-to-use
Catalog No. NC3074110 Supplier BIOLIFE SOLUTIONS LLC Supplier No. 210502
CryoStor CS10, 100 mL, pre-formulated, serum-free, protein-free, animal component-free, 10% DMSO, ready-to-use