Learn More
STEMCELL Technologies CryoStor CS10, Size: 1000 mL
SDP
Catalog No. 501974439
Encompass
Maximize post-thaw cell recovery and viability following cryopreservation at very low temperatures (-80°C to -196°C) with ready-to-use CryoStor CS10 medium. Serum-free, animal component-free, and cGMP-manufactured, this defined medium provides a safe, protective environment that is recommended for the cryopreservation of a variety of sensitive cell and sample types, including myeloma cell lines, human pluripotent stem cells, human peripheral blood, and more. Available in a variety of convenient formats, CryoStor CS10 is formulated with USP-grade components to minimize variability and contains 10% DMSO. Visit STEMCELL.com/cryostor-cs10 for more information.
Catalog No. 50-197-4439 Supplier STEMCELL Technologies Supplier No. 07940
Add to Cart
Add to Cart
Request a Quote
Maximize post-thaw cell recovery and viability following cryopreservation at very low temperatures (-80°C to -196°C) with ready-to-use CryoStor CS10 medium. Serum-free, animal component-free, and cGMP-manufactured, this defined medium provides a safe, protective environment that is recommended for the cryopreservation of a variety of sensitive cell and sample types, including myeloma cell lines, human pluripotent stem cells, human peripheral blood, and more. Available in a variety of convenient formats, CryoStor CS10 is formulated with USP-grade components to minimize variability and contains 10% DMSO. Visit STEMCELL.com/cryostor-cs10 for more information.