Learn More
BIOLIFE SOLUTIONS INC CryoStor CS2 Freeze Media, 100 mL
SDP
Catalog No. NC0991213
Encompass
CryoStor Pre-formulated with 2% DMSO. In some cell types, CryoStor CS2 has demonstrated biopreservation efficacy at or above the levels of competing commercial and in-house formulated freeze media, even in the presence of greatly reduced levels of DMSO.
This item is not returnable. View return policy
Catalog No. NC0991213 Supplier BIOLIFE SOLUTIONS INC Supplier No. 202102
CryoStor Pre-formulated with 2% DMSO. In some cell types, CryoStor CS2 has demonstrated biopreservation efficacy at or above the levels of competing commercial and in-house formulated freeze media, even in the presence of greatly reduced levels of DMSO.