Learn More
STEMCELL Technologies CryoStor CS2, Size: 100 mL
SDP
Catalog No. 501974433
Encompass
Maximize post-thaw cell recovery and viability following cryopreservation at low temperatures (-80°C to -196°C) with ready-to-use CryoStor CS2. Serum-free, animal component-free, and cGMP-manufactured, this defined medium provides a safe, protective environment for cells and tissues during freezing and thawing processes and storage. CryoStor CS2 is formulated with USP-grade components and contains 2% DMSO.
Catalog No. 50-197-4433 Supplier STEMCELL Technologies Supplier No. 07932
Maximize post-thaw cell recovery and viability following cryopreservation at low temperatures (-80°C to -196°C) with ready-to-use CryoStor CS2. Serum-free, animal component-free, and cGMP-manufactured, this defined medium provides a safe, protective environment for cells and tissues during freezing and thawing processes and storage. CryoStor CS2 is formulated with USP-grade components and contains 2% DMSO.