Learn More
STEMCELL Technologies CryoStor CS5, Size: 100 mL
SDP
Catalog No. NC1494931
Encompass
Maximize post-thaw cell recovery and viability following cryopreservation at low temperatures (-80°C to -196°C) with ready-to-use CryoStor CS5. Serum-free, animal component-free, and cGMP-manufactured, this defined medium provides a safe, protective environment for cells and tissues during freezing and thawing processes and storage. Available in a variety of convenient formats, CryoStor CS5 is formulated with USP-grade components and contains 5% DMSO. Visit STEMCELL.com for more information.
Catalog No. NC1494931 Supplier STEMCELL Technologies Supplier No. 07933
Maximize post-thaw cell recovery and viability following cryopreservation at low temperatures (-80°C to -196°C) with ready-to-use CryoStor CS5. Serum-free, animal component-free, and cGMP-manufactured, this defined medium provides a safe, protective environment for cells and tissues during freezing and thawing processes and storage. Available in a variety of convenient formats, CryoStor CS5 is formulated with USP-grade components and contains 5% DMSO. Visit STEMCELL.com for more information.