Learn More
BIOLIFE SOLUTIONS INC CryoStor CSB, 1000mL Bag
SDP
Catalog No. NC1650039
Encompass
CryoStor CSB is the intracellular-like base formulation for our series of cell-specific, optimized CryoStor preservation media. It is uniquely formulated to address the molecular-biological aspects of cells during the cryopreservation process. This enables a reduction in the level of cryopreservation-induced Delayed-Onset Cell Death and improvement in post-thaw cell viability and function.
This item is not returnable. View return policy
Catalog No. NC1650039 Supplier BIOLIFE SOLUTIONS INC Supplier No. 200210
Add to Cart
Add to Cart
Request a Quote
CryoStor CSB is the intracellular-like base formulation for our series of cell-specific, optimized CryoStor preservation media. It is uniquely formulated to address the molecular-biological aspects of cells during the cryopreservation process. This enables a reduction in the level of cryopreservation-induced Delayed-Onset Cell Death and improvement in post-thaw cell viability and function.