Learn More
Origen Biomedical CRYOSTORE EVA BAGS 500ML 24/CS
Catalog No. NC9089749
Encompass
  • Constructed from Ethyl Vinyl Acetate (EVA) film for durability
  • Nominal volume of 500 ml for efficient cryopreservation
  • Freeze volume range between 55 ml and 100 ml
  • Standard tubing set with two female luers with caps
  • Features hermetically sealed spike ports for secure closure
  • Ensures traceability with heat-welded lot numbers
  • Sterile and pyrogen-free fluid path, designed for single use
  • Packaged in cases of 24 bags for convenience
  • CE marked, FDA cleared, and ISO certified for compliance
  • Ideal for cryopreservation of blood components in clinical settings
  • Stable at ultra-low cryogenic temperatures down to -196°C
  • High flexibility, impact resistance, and puncture resistance for reliability
  • Low water vapor transmission for effective gas barrier properties
  • Designed with a corner-placed donor tube for easy de-bubbling and sealing
Catalog No. NC9089749 Supplier Origen Biomedical Supplier No. CS500
  • Constructed from Ethyl Vinyl Acetate (EVA) film for durability
  • Nominal volume of 500 ml for efficient cryopreservation
  • Freeze volume range between 55 ml and 100 ml
  • Standard tubing set with two female luers with caps
  • Features hermetically sealed spike ports for secure closure
  • Ensures traceability with heat-welded lot numbers
  • Sterile and pyrogen-free fluid path, designed for single use
  • Packaged in cases of 24 bags for convenience
  • CE marked, FDA cleared, and ISO certified for compliance
  • Ideal for cryopreservation of blood components in clinical settings
  • Stable at ultra-low cryogenic temperatures down to -196°C
  • High flexibility, impact resistance, and puncture resistance for reliability
  • Low water vapor transmission for effective gas barrier properties
  • Designed with a corner-placed donor tube for easy de-bubbling and sealing