Learn More
Origen Biomedical CRYOSTORE FLEX BAG 24/CS
Catalog No. NC2090168
Encompass
  • Durable construction from EVA material.
  • Independent sealing for individual 4-6 ml chambers post-thaw.
  • Bag fill/freeze volume of 20-30 ml per bag.
  • Bag length of 5.55 in.
  • Bag width of 3.32 in.
  • Includes "N" tube set for enhanced flexibility and safety.
  • Needle-free injection port for user convenience.
  • Manufactured under GMP guidelines for quality assurance.
  • CE marked and FDA 510(k) cleared for regulatory compliance.
  • Includes Certificate of Conformance (CoC).
  • Pouch of 4 bags for inner packaging.
  • Case quantity of 24 bags.
  • Compatible with CryoStore CS50 cassette.
This item is not returnable. View return policy
Catalog No. NC2090168 Supplier Origen Biomedical Supplier No. CS20N
  • Durable construction from EVA material.
  • Independent sealing for individual 4-6 ml chambers post-thaw.
  • Bag fill/freeze volume of 20-30 ml per bag.
  • Bag length of 5.55 in.
  • Bag width of 3.32 in.
  • Includes "N" tube set for enhanced flexibility and safety.
  • Needle-free injection port for user convenience.
  • Manufactured under GMP guidelines for quality assurance.
  • CE marked and FDA 510(k) cleared for regulatory compliance.
  • Includes Certificate of Conformance (CoC).
  • Pouch of 4 bags for inner packaging.
  • Case quantity of 24 bags.
  • Compatible with CryoStore CS50 cassette.