Learn More
Origen Biomedical CRYOSTORE FREEZING BAG 24 CS
Catalog No. NC0794639
Encompass
  • Constructed from durable EVA film
  • Hermetically sealed spike ports maintain sample integrity
  • N tube set configuration includes various caps and ports for sterile processing
  • Nominal freeze volume of 10-25 ml
  • Bag dimensions: 9.0 cm width, 7.3 cm length
  • Printed bag number and segments enhance traceability
  • Sterile; meant for single use only
  • FDA 510(k) cleared and CE marked
  • Manufactured under ISO 13485 and GMP guidelines
  • Biologically safe and free from phthalates, BPA, and latex
  • Packaged with 24 bags per case
Catalog No. NC0794639 Supplier Origen Biomedical Supplier No. CS25N
  • Constructed from durable EVA film
  • Hermetically sealed spike ports maintain sample integrity
  • N tube set configuration includes various caps and ports for sterile processing
  • Nominal freeze volume of 10-25 ml
  • Bag dimensions: 9.0 cm width, 7.3 cm length
  • Printed bag number and segments enhance traceability
  • Sterile; meant for single use only
  • FDA 510(k) cleared and CE marked
  • Manufactured under ISO 13485 and GMP guidelines
  • Biologically safe and free from phthalates, BPA, and latex
  • Packaged with 24 bags per case