Learn More
Research Products International Corp Cryostore Vial with Lip Seal, Flat Top Screw Cap Pre-Attached, Sterile, 2.0ml Capacity, 500 per Case
SDP
Catalog No. 50998593
Encompass
Perfect for cryogenic work, these vials come with seals in a flat top screw cap which provides a writing surface. Caps are loosely pre-attached to vials to ensure product sterility. Freestanding polypropylene vials feature white printed graduations and marking area.
Withstand -196°C temperatures
Leak-proof "O" ring screw caps
RNase and DNase free, sterile
Skirted
Catalog No. 50-998-593 Supplier Research Products International Corp Supplier No. 212408
Perfect for cryogenic work, these vials come with seals in a flat top screw cap which provides a writing surface. Caps are loosely pre-attached to vials to ensure product sterility. Freestanding polypropylene vials feature white printed graduations and marking area.
Withstand -196°C temperatures
Leak-proof "O" ring screw caps
RNase and DNase free, sterile
Skirted