Learn More
Alkali Scientific Cryogenic Storage Box, 25 place (5 x 5), Polycarbonate, with Pull-off Lid, (5.25 x 5.25 x 2.75"), 5/pk
SDP
Catalog No. 502336104
Encompass_Preferred
Ultra-low temperature Cryogenic Storage Boxes are designed to hold 1.2mL to 2.0mL cryogenic storage vials. An excellent and longlasting replacement for cardboard boxes, they are fully autoclavable and withstand temperatures ranging from -196° to +121°C. They are compatible with all standard 5.25 inch (133mm) freezer racks. Clear lids are hinged and have printed indexing for sample identification purposes and include drainage slots for LN2.
Catalog No. 50-233-6104 Supplier Alkali Scientific Supplier No. C2581
Ultra-low temperature Cryogenic Storage Boxes are designed to hold 1.2mL to 2.0mL cryogenic storage vials. An excellent and longlasting replacement for cardboard boxes, they are fully autoclavable and withstand temperatures ranging from -196° to +121°C. They are compatible with all standard 5.25 inch (133mm) freezer racks. Clear lids are hinged and have printed indexing for sample identification purposes and include drainage slots for LN2.