Learn More
Alkali Scientific Cryostorage Box, 81 Place (9X9), Polycarbonate, with Hinged Lid (5.25 X 5.25 X 2 in), 5/cs
SDP
Catalog No. 502335899
Encompass_Preferred
Ultra-low temperature storage boxes are designed to hold 1.2mL to 2.0mL cryogenic storage vials. An excellent and long-lasting replacement for cardboard boxes, they are fully autoclavable and withstand temperatures ranging from -196° to +121°C. they are compatible with all standard 5.25 inch (133mm) freezer racks. Clear Lids are Hinged and have printed indexing for sample identification purposes.
Catalog No. 50-233-5899 Supplier Alkali Scientific Supplier No. MT2021
Ultra-low temperature storage boxes are designed to hold 1.2mL to 2.0mL cryogenic storage vials. An excellent and long-lasting replacement for cardboard boxes, they are fully autoclavable and withstand temperatures ranging from -196° to +121°C. they are compatible with all standard 5.25 inch (133mm) freezer racks. Clear Lids are Hinged and have printed indexing for sample identification purposes.