Learn More
Electron Microscopy Sciences CryoSTUCK Thermal Transfer Labels | 25.4 x 15.9mm +35mm wrap | Perf: Yes + Notch
SDP
Catalog No. 501930305
Encompass_Preferred
These cryogenic labels have a clear overlap area and can be applied directly to frozen tubes and cryovials, even those difficult to label in deep-freeze conditions. Frozen vials can be labeled seconds after removal from cryogenic storage and can be placed back into liquid nitrogen or a freezer for further conservation. They are designed for long-term preservation in liquid nitrogen freezers and dry ice. They can withstand temperatures up to +100 deg C and the extra permanent adhesive allows for application to frozen containers at temperatures as low as -80 deg C. Temperature range is from -196 deg C to +100 deg C.The labels are compatible with most brands of thermal transfer printers. For proper adherence to the frozen surface, we recommend applying the CryoSTUCK labels with an overlap of the transparent part of at least 0.25".Size: 1" x 5/8" + 1.375" Color: White, Core: 1", Labels across: 1 .
Catalog No. 50-193-0305 Supplier Electron Microscopy Sciences Supplier No. 7742212W
These cryogenic labels have a clear overlap area and can be applied directly to frozen tubes and cryovials, even those difficult to label in deep-freeze conditions. Frozen vials can be labeled seconds after removal from cryogenic storage and can be placed back into liquid nitrogen or a freezer for further conservation. They are designed for long-term preservation in liquid nitrogen freezers and dry ice. They can withstand temperatures up to +100 deg C and the extra permanent adhesive allows for application to frozen containers at temperatures as low as -80 deg C. Temperature range is from -196 deg C to +100 deg C.The labels are compatible with most brands of thermal transfer printers. For proper adherence to the frozen surface, we recommend applying the CryoSTUCK labels with an overlap of the transparent part of at least 0.25".Size: 1" x 5/8" + 1.375" Color: White, Core: 1", Labels across: 1 .