Learn More
Med Vet International Cool Renewal™ Complete Cryosurgical Kit, J1401
Catalog No. 5000495447
Encompass_Preferred
Jorvets Cool Renewal Cryosurgical kit is a convenient alternative to liquid nitrogen. It has a spray canister that freezes at -94 degrees farenheit, offers minimal patient discomfort and is slef anesthetizing. This kit is portable and allows for a 2 minute procedure. This cryosurgical kit includes: 2 - 170ml canisters of cryogen-130 freezes 2 - Extender Tubes Assorted pack of 60 disposable double ended foamm tipped applicators Assorted pack of 50 disposable plastic isolation funnels 10 - Disposable skin tag tweezers Instructions for use Patient instructions tear pad Applicator practice pad Magentic packaging for component storage
Catalog No. 50-004-95447 Supplier Med Vet International Supplier No. JOR1401
Jorvets Cool Renewal Cryosurgical kit is a convenient alternative to liquid nitrogen. It has a spray canister that freezes at -94 degrees farenheit, offers minimal patient discomfort and is slef anesthetizing. This kit is portable and allows for a 2 minute procedure. This cryosurgical kit includes: 2 - 170ml canisters of cryogen-130 freezes 2 - Extender Tubes Assorted pack of 60 disposable double ended foamm tipped applicators Assorted pack of 50 disposable plastic isolation funnels 10 - Disposable skin tag tweezers Instructions for use Patient instructions tear pad Applicator practice pad Magentic packaging for component storage