Learn More
MVE BIOLOGICAL SOLUTIONS MVE CryoSystem 750, 750 Vial Capacity, Efficient Liquid Nitrogen Use, 47.4 L Capacity
Catalog No. NC2741859
Encompass_Preferred
The MVE CryoSystem 750 is an advanced cryogenic sample storage system designed for efficient preservation of biological samples. With a 750 vial capacity, lightweight aluminum construction, and exceptional thermal performance, it ensures reliable storage with low liquid nitrogen use. Its tamper-proof lid and intelligent canister design enhance security and ease of access, distinguishing it from other storage solutions.
Catalog No. NC2741859 Supplier MVE BIOLOGICAL SOLUTIONS Supplier No. 11886450
The MVE CryoSystem 750 is an advanced cryogenic sample storage system designed for efficient preservation of biological samples. With a 750 vial capacity, lightweight aluminum construction, and exceptional thermal performance, it ensures reliable storage with low liquid nitrogen use. Its tamper-proof lid and intelligent canister design enhance security and ease of access, distinguishing it from other storage solutions.