Learn More
Research Products International Corp Cryo-Tags Labels, Laser Sheet, 2.63 x 1 Inch, Yellow, 600 per Package
SDP
Supplier:  Research Products International Corp 247138Y
Encompass
Convenient Cryo-Tags offer superior temperature and chemical resistance. Durable labels withstand cryogenic liquid nitrogen storage, boiling water baths, solvents, caustic solutions and other laboratory chemicals. Cryo-Tags fit bottles, flasks, vials, cryoboxes and more. Chemically inert poly vinyl labels adhere to all polypropylene and polyethylene surfaces without cracking, peeling or degrading and can be marked with a regular pen or marker.
Catalog No. 50-998-673