Learn More
Andwin Scientific CRYOTAGS WH 38X19 1200/PK
SDP
Supplier:  Andwin Scientific 77564W
Encompass_Preferred
The 1.50" L x 0.5" W Cryo-Babies labels can withstand most organic solvents, caustic agents, temperatures from -196C to 100C. This product can still be utilized while under oven, autoclaving, or extreme freezer temperatures. The labels can also be used on several surfaces like plastics, glass, and metals without cracking, peeling, or degrading. This product is sold in the color of white.
Catalog No. 50-727-680