Learn More
GA International Cryogenic Tape for Frozen Surfaces 0.5" x 50' / 12.7mm x 15m Blue 1" / 25.4mm From -196°C to +100°C (-321°F to +212°F) / Minimum Application Temperature: -80°C (-112°F)
Catalog No. NC1589371
Encompass_Preferred
Cryogenic tape for the identification of already frozen containers such as cryo boxes, cardboard and plastic freezer boxes, as well as tubes and bottles which are difficult to label under deep-freeze conditions (-80°C/-112°F). Ideal for long-term cryo-preservation in liquid and vapor phase liquid nitrogen (-196°C/-321°F), laboratory freezers, and dry ice transportation. These cryo tapes can be inscribed using our permanent cryo markers and are available in an assortment of colors for easy identification and color coding.
Catalog No. NC1589371 Supplier GA International Supplier No. TFS13C150BL
Cryogenic tape for the identification of already frozen containers such as cryo boxes, cardboard and plastic freezer boxes, as well as tubes and bottles which are difficult to label under deep-freeze conditions (-80°C/-112°F). Ideal for long-term cryo-preservation in liquid and vapor phase liquid nitrogen (-196°C/-321°F), laboratory freezers, and dry ice transportation. These cryo tapes can be inscribed using our permanent cryo markers and are available in an assortment of colors for easy identification and color coding.