Learn More
Research Products International Corp Cryotube Storage Box, Polypropylene, 81 Tube Capacity, 4 per Case
SDP
Catalog No. 50212261
Encompass
Storage box is designed for durable and convenient storage of up to 81 (1.2 and 2.0 ml) cryotubes. Constructed of rugged polypropylene, a frosted writing surface on the face of the lid permits quick identification of the box. Designed for use with either mechanically refrigerated or vapor phase liquid nitrogen systems.
Catalog No. 50-212-261 Supplier Research Products International Corp Supplier No. 142500
Storage box is designed for durable and convenient storage of up to 81 (1.2 and 2.0 ml) cryotubes. Constructed of rugged polypropylene, a frosted writing surface on the face of the lid permits quick identification of the box. Designed for use with either mechanically refrigerated or vapor phase liquid nitrogen systems.