Learn More
CAPTIVATE BIO LLC CRYOVATE FD Media
Catalog No. 5000876338
Encompass_Preferred
Captivate Bio s CRYOVATE FD Freezing Medium is a traditional easy-to-use cryopreservation media for various types of animal and human adherent or suspension cells CRYOVATE FD contains FBS and DMSO and is designed to protect and preserve cells during long-term storage Manufactured in the US
Catalog No. 50-008-76338 Supplier CAPTIVATE BIO LLC Supplier No. CRY004
Captivate Bio s CRYOVATE FD Freezing Medium is a traditional easy-to-use cryopreservation media for various types of animal and human adherent or suspension cells CRYOVATE FD contains FBS and DMSO and is designed to protect and preserve cells during long-term storage Manufactured in the US