Learn More
CAPTIVATE BIO LLC CRYOVATE SF Media
Catalog No. 5000876350
Encompass_Preferred
Captivate Bio s CRYOVATE SF Freezing Medium is a sterile ready-to-use serum-free cryopreservation medium that allows for the optimal storage of human and animal cells This formulation contains DMSO and other cryoprotectants to ensure cell integrity and viability
Catalog No. 50-008-76350 Supplier CAPTIVATE BIO LLC Supplier No. CRY014
Captivate Bio s CRYOVATE SF Freezing Medium is a sterile ready-to-use serum-free cryopreservation medium that allows for the optimal storage of human and animal cells This formulation contains DMSO and other cryoprotectants to ensure cell integrity and viability