Learn More
Cole-Parmer Argos Technologies PolarSafe Sterile 2D Cryovials, 2 mL, Skirted-Bottom, External Thread; 50/Pk
Catalog No. 501960592 Shop All Cole-Parmer Products
Encompass_Preferred
The PolarSafe cryogenic 2D vials are available in 2 and 5 mL sizes with internally or externally threaded caps. They are ideal for rapid processing of biological samples. The embedded Datamatrix code at the bottom of each vial, along with the corresponding Code 128 barcode on the body of the vial saves time and provides accurate organization, and tracking of valuable samples. The 2D vials can be safely stored in polycarbonate boxes that can be purchase separately. The storage boxes also features the 2D Datamatrix code and Code 128 barcode to provide automatic identification of both the box and stored vials.
Catalog No. 50-196-0592 Supplier Cole-Parmer Supplier No. 0439580
The PolarSafe cryogenic 2D vials are available in 2 and 5 mL sizes with internally or externally threaded caps. They are ideal for rapid processing of biological samples. The embedded Datamatrix code at the bottom of each vial, along with the corresponding Code 128 barcode on the body of the vial saves time and provides accurate organization, and tracking of valuable samples. The 2D vials can be safely stored in polycarbonate boxes that can be purchase separately. The storage boxes also features the 2D Datamatrix code and Code 128 barcode to provide automatic identification of both the box and stored vials.