Learn More
CELLTREAT Scientific Products Red Cap Insert for CF Cryogenic Vials, Non-sterile
SDP
Catalog No. 501149088
Encompass_Preferred
CELLTREAT Scientific Products Red Cap insert for Freeze Cryogenic Vials. Easily color-code samples with colored cap inserts. CELLTREAT Freeze Cap Inserts are compatible with all CELLTREAT Freeze Cryogenic Vials. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged non-sterile, bulk packed 500/re-sealable bag, 500/case.
Catalog No. 50-114-9088 Supplier CELLTREAT Scientific Products Supplier No. 229930
CELLTREAT Scientific Products Red Cap insert for Freeze Cryogenic Vials. Easily color-code samples with colored cap inserts. CELLTREAT Freeze Cap Inserts are compatible with all CELLTREAT Freeze Cryogenic Vials. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged non-sterile, bulk packed 500/re-sealable bag, 500/case.