Learn More
Mitegen Cryovial Clamp - Curved, for Manipulating Cryovials
SDP
Catalog No. 501492128
Encompass_Preferred
Hemostat tool for manipulating cryovials in liquid nitrogen environments.
Catalog No. 50-149-2128 Supplier Mitegen Supplier No. 1013195
Hemostat tool for manipulating cryovials in liquid nitrogen environments.