Learn More
Mitegen Cryovial Clamp Set
SDP
Catalog No. 501492126
Encompass_Preferred
Hemostat tool for manipulating cryovials in liquid nitrogen environments. Kit contains one straight and one curved model.
Catalog No. 50-149-2126 Supplier Mitegen Supplier No. 1013193
Hemostat tool for manipulating cryovials in liquid nitrogen environments. Kit contains one straight and one curved model.