Learn More
Chemglass Life Sciences Cool Block, Aluminum, 32 Position, 1.5mL and 2.0mL Cryovials
SDP
Catalog No. 501532369
Encompass
Cool Block, Aluminum, 32 Position, 1.5mL and 2.0mL Cryovials. Block is made from solid aluminum and is specially designed for cryovials with a locking mechanism to engage the slots on the bottom of the vials. This unique feature allows for single handed operation to open and close the cap of the tube. Dimensions are 6.5 x 3 x 1.5
Catalog No. 50-153-2369 Supplier Chemglass Life Sciences Supplier No. CLS477401
Cool Block, Aluminum, 32 Position, 1.5mL and 2.0mL Cryovials. Block is made from solid aluminum and is specially designed for cryovials with a locking mechanism to engage the slots on the bottom of the vials. This unique feature allows for single handed operation to open and close the cap of the tube. Dimensions are 6.5 x 3 x 1.5