Learn More
Electron Microscopy Sciences CryoVial CoolSafe System for 2.0 ml cryovials
SDP
Catalog No. 5034162
Encompass_Preferred
The Cooling chamber is suitable for one-hand operation with locking base holding vial in place as cap is removed.
It fits most major cryogenic vial brands with universal design.
Preserve cells by minimizing cell loss from exothermic reactions by preparing samples inside chambers.
Each aluminum CryoVial Cooling Chamber surrounds tubes to guarantee sustained cold temperatures.
Each Chamber is anodized clear to protect from rust and corrosion ensuring longevity.
Use with Cool Bricks to maintain a constant temperature (1 degree C or –10 degree C) for up to 10 hours.
Complete CryoVial CoolSafe System includes 1 CoolSafe box with lid, 1 CryoVial Cooling Chamber, and 2 Cool Bricks (1 degree C & -10 degree C). Includes: 1 CoolSafe box with lid, 1 CryoVial Cooling Chamber and 2 Cool Bricks (1 deg C & -10 deg C).
Use: 2.0 ml cryovials
Catalog No. 50-341-62 Supplier Electron Microscopy Sciences Supplier No. 6195632
The Cooling chamber is suitable for one-hand operation with locking base holding vial in place as cap is removed.
It fits most major cryogenic vial brands with universal design.
Preserve cells by minimizing cell loss from exothermic reactions by preparing samples inside chambers.
Each aluminum CryoVial Cooling Chamber surrounds tubes to guarantee sustained cold temperatures.
Each Chamber is anodized clear to protect from rust and corrosion ensuring longevity.
Use with Cool Bricks to maintain a constant temperature (1 degree C or –10 degree C) for up to 10 hours.
Complete CryoVial CoolSafe System includes 1 CoolSafe box with lid, 1 CryoVial Cooling Chamber, and 2 Cool Bricks (1 degree C & -10 degree C). Includes: 1 CoolSafe box with lid, 1 CryoVial Cooling Chamber and 2 Cool Bricks (1 deg C & -10 deg C).
Use: 2.0 ml cryovials