Learn More
Alkali Scientific ASI™ Cryovial Rack with numeric labeling, 5x10 (50 holes) Blue,Green, Red, PP, autoclavable, 10/Pack
SDP
Catalog No. 503026705
Encompass_Preferred
Numeric labeling 3 colors: Blue, Green, Red Autoclavable Material: PP Description: 5 x 10 (50 Holes) Packaging: 10 Made of polypropylene This handy autoclavable rack can hold up to 50 cryogenic vials. Now with one hand, you can easily unscrew a cryovial closure. Thanks to an innovative universal locking system, the vials will securely lock in each well and will not turn. Each position is identified with an alphanumeric index. Strong handles make it easy and safe to carry. It is supported by anti-skid rubber feet. The rack is compact and stackable. Available in three attractive colors. Size: 10 cm x 20 cm x 25 mm H (4 x 8 x 1 in. H)
Catalog No. 50-302-6705 Supplier Alkali Scientific Supplier No. VCRK
Numeric labeling 3 colors: Blue, Green, Red Autoclavable Material: PP Description: 5 x 10 (50 Holes) Packaging: 10 Made of polypropylene This handy autoclavable rack can hold up to 50 cryogenic vials. Now with one hand, you can easily unscrew a cryovial closure. Thanks to an innovative universal locking system, the vials will securely lock in each well and will not turn. Each position is identified with an alphanumeric index. Strong handles make it easy and safe to carry. It is supported by anti-skid rubber feet. The rack is compact and stackable. Available in three attractive colors. Size: 10 cm x 20 cm x 25 mm H (4 x 8 x 1 in. H)