Learn More
Electron Microscopy Sciences CryoVial Series T309-2, 2 ml, Round-Bottom, 12.5 x 48 mm
SDP
Catalog No. 50364428
Encompass_Preferred
Specially Formulated Polypropylene, is certified Rnase-free, Dnase-free, Pyrogen-free, and DNA-free. Series T309 is a lip-seal design tube with external threads. Series T310 has a silicone washer seal and external threads. Series T311 has a silicone washer seal with internal threads. Lip-seal design, external threads.
Volume: 2 ml
Base: Round-Bottom
Size: 12.5 x 48 mm
Catalog No. 50-364-428 Supplier Electron Microscopy Sciences Supplier No. 6180002
Specially Formulated Polypropylene, is certified Rnase-free, Dnase-free, Pyrogen-free, and DNA-free. Series T309 is a lip-seal design tube with external threads. Series T310 has a silicone washer seal and external threads. Series T311 has a silicone washer seal with internal threads. Lip-seal design, external threads.
Volume: 2 ml
Base: Round-Bottom
Size: 12.5 x 48 mm