Learn More
Electron Microscopy Sciences CryoVial Series T309-4A, 4 ml, Self-Standing, 12.5 x 78 mm, Case of 1000 (10 packs of 100 ea)
SDP
Catalog No. 501899026
Encompass_Preferred
Specially Formulated Polypropylene, is certified Rnase-free, Dnase-free, Pyrogen-free, and DNA-free. Series T309 is a lip-seal design tube with external threads. Series T310 has a silicone washer seal and external threads. Series T311 has a silicone washer seal with internal threads. Lip-seal design, external threads.
Volume: 4 ml
Base: Self-Standing
Size: 12.5 x 78 mm
Catalog No. 50-189-9026 Supplier Electron Microscopy Sciences Supplier No. 618004A
Specially Formulated Polypropylene, is certified Rnase-free, Dnase-free, Pyrogen-free, and DNA-free. Series T309 is a lip-seal design tube with external threads. Series T310 has a silicone washer seal and external threads. Series T311 has a silicone washer seal with internal threads. Lip-seal design, external threads.
Volume: 4 ml
Base: Self-Standing
Size: 12.5 x 78 mm