Learn More
Electron Microscopy Sciences CryoVial Series T309, 10 ml, Self-Standing, 17 x 84 mm
SDP
Catalog No. 501927749
Encompass_Preferred
Specially Formulated Polypropylene, is certified Rnase-free, Dnase-free, Pyrogen-free, and DNA-free. Series T309 is a lip-seal design tube with external threads. Series T310 has a silicone washer seal and external threads. Series T311 has a silicone washer seal with internal threads. Silicone washer seal, external threads.
Volume: 10 ml
Base: Self-Standing
Size: 17 x 84 mm
This item is not returnable. View return policy
Catalog No. 50-192-7749 Supplier Electron Microscopy Sciences Supplier No. 6180110A100
Specially Formulated Polypropylene, is certified Rnase-free, Dnase-free, Pyrogen-free, and DNA-free. Series T309 is a lip-seal design tube with external threads. Series T310 has a silicone washer seal and external threads. Series T311 has a silicone washer seal with internal threads. Silicone washer seal, external threads.
Volume: 10 ml
Base: Self-Standing
Size: 17 x 84 mm