Learn More
Electron Microscopy Sciences CryoVial Series T310-2, 2 ml, Round-Bottom, 12.5 x 48 mm
SDP
Catalog No. 501899027
Encompass_Preferred
Specially Formulated Polypropylene, is certified Rnase-free, Dnase-free, Pyrogen-free, and DNA-free. Series T309 is a lip-seal design tube with external threads. Series T310 has a silicone washer seal and external threads. Series T311 has a silicone washer seal with internal threads. Silicone washer seal, external threads.
Volume: 2 ml
Base: Round-Bottom
Size: 12.5 x 48 mm
Catalog No. 50-189-9027 Supplier Electron Microscopy Sciences Supplier No. 6180102
Specially Formulated Polypropylene, is certified Rnase-free, Dnase-free, Pyrogen-free, and DNA-free. Series T309 is a lip-seal design tube with external threads. Series T310 has a silicone washer seal and external threads. Series T311 has a silicone washer seal with internal threads. Silicone washer seal, external threads.
Volume: 2 ml
Base: Round-Bottom
Size: 12.5 x 48 mm