Learn More
Electron Microscopy Sciences CryoVial Series T309, 3 ml, Self-Standing, 12.5 x 72 mm
SDP
Catalog No. 501927752
Encompass_Preferred
Specially Formulated Polypropylene, is certified Rnase-free, Dnase-free, Pyrogen-free, and DNA-free. Series T309 is a lip-seal design tube with external threads. Series T310 has a silicone washer seal and external threads. Series T311 has a silicone washer seal with internal threads. Silicone washer seal, external threads.
Volume: 3 ml
Base: Self-Standing
Size: 12.5 x 72 mm
Catalog No. 50-192-7752 Supplier Electron Microscopy Sciences Supplier No. 618013A100
Specially Formulated Polypropylene, is certified Rnase-free, Dnase-free, Pyrogen-free, and DNA-free. Series T309 is a lip-seal design tube with external threads. Series T310 has a silicone washer seal and external threads. Series T311 has a silicone washer seal with internal threads. Silicone washer seal, external threads.
Volume: 3 ml
Base: Self-Standing
Size: 12.5 x 72 mm