Learn More
Electron Microscopy Sciences CryoVial Series T309, 4 ml, Round-Bottom, 12.5 x 72 mm
SDP
Catalog No. 501927785
Encompass_Preferred
Specially Formulated Polypropylene, is certified Rnase-free, Dnase-free, Pyrogen-free, and DNA-free. Series T309 is a lip-seal design tube with external threads. Series T310 has a silicone washer seal and external threads. Series T311 has a silicone washer seal with internal threads. Silicone washer seal, internal threads.
Volume: 4 ml
Base: Round-Bottom
Size: 12.5 x 72 mm
Catalog No. 50-192-7785 Supplier Electron Microscopy Sciences Supplier No. 618024A100
Specially Formulated Polypropylene, is certified Rnase-free, Dnase-free, Pyrogen-free, and DNA-free. Series T309 is a lip-seal design tube with external threads. Series T310 has a silicone washer seal and external threads. Series T311 has a silicone washer seal with internal threads. Silicone washer seal, internal threads.
Volume: 4 ml
Base: Round-Bottom
Size: 12.5 x 72 mm