Learn More
Mitegen CryoVial Tong, Straight Design, For Safe & Secure Grasping, Positioning & Transfer of Magnetic CryoVials
SDP
Catalog No. 501187414
Encompass
This CryoVial Tong with a straight design provides an essential solution for the safe, secure, and precise manipulation of CryoVials in various laboratory settings. These tongs are compatible with cryovials such as SKUs CV-1-100, CV-1-200, CV-1-50 and CV-1-500. Engineered for optimal control, this tong allows users to reliably grasp, position, and transfer magnetic CryoVials, particularly in demanding cryogenic environments such as liquid nitrogen (LN2) dewars or ultra-low temperature freezers. The robust straight design ensures a firm hold on vials, minimizing the risk of accidental drops, thermal shock, or contamination of valuable biological samples during critical handling procedures in crystallography workflows. By enabling hands-free operation and controlled movement, this tong streamlines laboratory efficiency and enhances safety in cryo-preservation and sample management.
Catalog No. 50-118-7414 Supplier Mitegen Supplier No. MR1013194
This CryoVial Tong with a straight design provides an essential solution for the safe, secure, and precise manipulation of CryoVials in various laboratory settings. These tongs are compatible with cryovials such as SKUs CV-1-100, CV-1-200, CV-1-50 and CV-1-500. Engineered for optimal control, this tong allows users to reliably grasp, position, and transfer magnetic CryoVials, particularly in demanding cryogenic environments such as liquid nitrogen (LN2) dewars or ultra-low temperature freezers. The robust straight design ensures a firm hold on vials, minimizing the risk of accidental drops, thermal shock, or contamination of valuable biological samples during critical handling procedures in crystallography workflows. By enabling hands-free operation and controlled movement, this tong streamlines laboratory efficiency and enhances safety in cryo-preservation and sample management.