Learn More
Mitegen Magnetic CryoVials, SPINE Standard, For Secure Cryogenic Sample Storage & Handling, 500 Vials/Pack
SDP
Catalog No. 501939455
Encompass
Magnetic CryoVials provide a secure and efficient solution for cryogenic sample storage and handling in diverse laboratory environments. Designed to adhere to the widely recognized SPINE Standard, these vials ensure broad compatibility with common cryo-storage systems, racks, and accessories, including magnetic canes. The integrated magnetic feature allows for effortless and precise manipulation of vials, minimizing direct contact with samples and reducing the risk of accidental drops or contamination during transfer in liquid nitrogen (LN2) dewars or bulk storage tanks. This enhances user safety and streamlines workflow, particularly during high-throughput operations. This is a 500 Vials/Pack. Cryovials can be easily handled with Cryovial tongs which are sold seperately see SKUs M-R-1013193 and M-R-1013194.
Catalog No. 50-193-9455 Supplier Mitegen Supplier No. CV1500
Magnetic CryoVials provide a secure and efficient solution for cryogenic sample storage and handling in diverse laboratory environments. Designed to adhere to the widely recognized SPINE Standard, these vials ensure broad compatibility with common cryo-storage systems, racks, and accessories, including magnetic canes. The integrated magnetic feature allows for effortless and precise manipulation of vials, minimizing direct contact with samples and reducing the risk of accidental drops or contamination during transfer in liquid nitrogen (LN2) dewars or bulk storage tanks. This enhances user safety and streamlines workflow, particularly during high-throughput operations. This is a 500 Vials/Pack. Cryovials can be easily handled with Cryovial tongs which are sold seperately see SKUs M-R-1013193 and M-R-1013194.