Learn More
Thermo Scientific™ Cryoware Labels
GSA_VAGREENER_CHOICE
Catalog No. 033377G Shop All Thermo Scientific Products
Change view
Click to view available options
Dimensions (L x W):
25 x 50 mm
Catalog No. Dimensions (L x W)
03-337-7G 25 x 50 mm
1 options
Catalog No. 03-337-7G Supplier Thermo Scientific™ Supplier No. 50400002
  • $295.50
    $258.65 / Pack of 200
    Save $36.85
    12% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $1,905.00
    $1,668.65 / Case of 10 PK
    Save $236.35
    12% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Easily and permanently identify samples and storage boxes with Thermo Scientific™ Cryoware Labels.
  • 25 x 50mm cloth labels
  • Accept markers or ballpoint pen
  • Specifically designed for use at ultra-low temperatures
  • Adhere to plastic and cardboard without shrinking or peeling
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
Adhesive Type Cold-temperature
Color White
Material Nylon Cloth
Sterility Non-sterile
Latex Free Yes
Dimensions (L x W) 25 x 50 mm
Voltage 0/1 V