Learn More
Medchemexpress LLC Cryptand 2.2.2 | 23978-09-8 | ≥98.0% | 376.49 | 250 MG
SDP
Catalog No. 5000451577
Encompass_Preferred
Cryptand 2.2.2 is a metal ion chelating agent that can selectively form complexes with various metal ions. At room temperature, it functions as a host molecule, utilizing a chelation effect to selectively bind desired ions. This product is useful in the preparation of nanoparticles and transition metal compounds.
  • Selectively binds desired metal ions such as Zn2+, Co2+, Ni2+, and Cu2+
  • Used in the preparation of nanoparticles
  • Used in the preparation of transition metal compounds
Catalog No. 50-004-51577 Supplier Medchemexpress LLC Supplier No. 250MGHYW011060
Cryptand 2.2.2 is a metal ion chelating agent that can selectively form complexes with various metal ions. At room temperature, it functions as a host molecule, utilizing a chelation effect to selectively bind desired ions. This product is useful in the preparation of nanoparticles and transition metal compounds.
  • Selectively binds desired metal ions such as Zn2+, Co2+, Ni2+, and Cu2+
  • Used in the preparation of nanoparticles
  • Used in the preparation of transition metal compounds