Learn More
Medchemexpress LLC Cryptochlorogenic acid (4-Caffeoylquinic acid) | 905-99-7 | 354.31 | 25 MG
SDP
Catalog No. 5000390155
Encompass_Preferred
Cryptochlorogenic acid (4-Caffeoylquinic acid) is a naturally occurring phenolic acid compound that is orally effective. It exhibits anti-inflammatory, antioxidant, and anti-cardiac hypertrophy effects. The compound can alleviate physiological stress responses by regulating proinflammatory factor expression, inhibiting NF-κB activity, promoting Nrf2 nuclear transfer, and regulating the PI3Kα/Akt/mTOR/HIF-1α signaling pathway.
  • Oral effectiveness
  • Anti-inflammatory effects
  • Antioxidant effects
  • Anti-cardiac hypertrophy effects
  • Regulates proinflammatory factor expression
  • Inhibits NF-κB activity
  • Promotes Nrf2 nuclear transfer
  • Regulates PI3Kα/Akt/mTOR/HIF-1α signaling pathway
Catalog No. 50-003-90155 Supplier Medchemexpress LLC Supplier No. 25MGHYN0787
Cryptochlorogenic acid (4-Caffeoylquinic acid) is a naturally occurring phenolic acid compound that is orally effective. It exhibits anti-inflammatory, antioxidant, and anti-cardiac hypertrophy effects. The compound can alleviate physiological stress responses by regulating proinflammatory factor expression, inhibiting NF-κB activity, promoting Nrf2 nuclear transfer, and regulating the PI3Kα/Akt/mTOR/HIF-1α signaling pathway.
  • Oral effectiveness
  • Anti-inflammatory effects
  • Antioxidant effects
  • Anti-cardiac hypertrophy effects
  • Regulates proinflammatory factor expression
  • Inhibits NF-κB activity
  • Promotes Nrf2 nuclear transfer
  • Regulates PI3Kα/Akt/mTOR/HIF-1α signaling pathway