Learn More
Cayman Chemical CryptochlorogenIc AcId 50mg
SDP
Catalog No. 502932736
Encompass_Preferred
A phenolic acid with diverse biological activities; inhibits RLAR in vitro (IC50 = 11.13 μM); inhibits the growth of S. aureus (MIC = 0.5 mg/ml); scavenges DPPH free radicals in a cell-free assay (IC50 = 5.26 μg/ml); induces degranulation of RBL-2H3 mast cells; increases plasma serotonin and β-hexosaminidase levels in guinea pigs
Catalog No. 50-293-2736 Supplier Cayman Chemical Supplier No. 2509850 MG
A phenolic acid with diverse biological activities; inhibits RLAR in vitro (IC50 = 11.13 μM); inhibits the growth of S. aureus (MIC = 0.5 mg/ml); scavenges DPPH free radicals in a cell-free assay (IC50 = 5.26 μg/ml); induces degranulation of RBL-2H3 mast cells; increases plasma serotonin and β-hexosaminidase levels in guinea pigs