Learn More
Medchemexpress LLC Cryptochlorogenic acid | 905-99-7 | 354.31 | 1 ML
SDP
Catalog No. 5000417039
Encompass_Preferred
Cryptochlorogenic acid is a naturally occurring phenolic acid compound with oral effectiveness, along with anti-inflammatory, antioxidant, and anti-cardiac hypertrophy effects. It functions by alleviating physiological stress responses by regulating proinflammatory factor expression, inhibiting NF-κB activity, promoting Nrf2 nuclear transfer, and regulating the PI3Kα/Akt/mTOR/HIF-1α signaling pathway. Intended for research use only.
  • Oral effectiveness
  • Anti-inflammatory and antioxidant effects
  • Anti-cardiac hypertrophy effects
  • Regulates proinflammatory factor expression
  • Inhibits NF-κB activity
  • Promotes Nrf2 nuclear transfer
  • Regulates PI3Kα/Akt/mTOR/HIF-1α signaling pathway
  • Low toxicity in RAW264.7 cells at specific concentrations
  • Reduces myocardial hypertrophy in H9c2 cells
Catalog No. 50-004-17039 Supplier Medchemexpress LLC Supplier No. HYN078710MM/1ML
Cryptochlorogenic acid is a naturally occurring phenolic acid compound with oral effectiveness, along with anti-inflammatory, antioxidant, and anti-cardiac hypertrophy effects. It functions by alleviating physiological stress responses by regulating proinflammatory factor expression, inhibiting NF-κB activity, promoting Nrf2 nuclear transfer, and regulating the PI3Kα/Akt/mTOR/HIF-1α signaling pathway. Intended for research use only.
  • Oral effectiveness
  • Anti-inflammatory and antioxidant effects
  • Anti-cardiac hypertrophy effects
  • Regulates proinflammatory factor expression
  • Inhibits NF-κB activity
  • Promotes Nrf2 nuclear transfer
  • Regulates PI3Kα/Akt/mTOR/HIF-1α signaling pathway
  • Low toxicity in RAW264.7 cells at specific concentrations
  • Reduces myocardial hypertrophy in H9c2 cells