Learn More
Medchemexpress LLC Cryptochlorogenic acid | 905-99-7 | MFCD10566638 | 354.31 | 100 MG
SDP
Catalog No. 5000416987
Encompass_Preferred
Cryptochlorogenic acid is a naturally occurring phenolic acid compound with oral effectiveness, anti-inflammatory, antioxidant, and anti-cardiac hypertrophy effects. It alleviates physiological stress responses by regulating proinflammatory factor expression, inhibiting NF-κB activity, promoting Nrf2 nuclear transfer, and regulating the PI3Kα/Akt/mTOR/HIF-1α signaling pathway.
  • Naturally occurring phenolic acid compound
  • Oral effectiveness
  • Anti-inflammatory
  • Antioxidant
  • Anti-cardiac hypertrophy effects
  • Alleviates LPS and ISO by regulating proinflammatory factor expression
  • Inhibits NF-κB activity
  • Promotes Nrf2 nuclear transfer
  • Regulates PI3Kα/Akt/mTOR/HIF-1α signaling pathway
  • Solid appearance, white to off-white color
  • Soluble in DMSO (50 mg/mL)
Catalog No. 50-004-16987 Supplier Medchemexpress LLC Supplier No. HYN0787100MG
Add to Cart
Add to Cart
Cryptochlorogenic acid is a naturally occurring phenolic acid compound with oral effectiveness, anti-inflammatory, antioxidant, and anti-cardiac hypertrophy effects. It alleviates physiological stress responses by regulating proinflammatory factor expression, inhibiting NF-κB activity, promoting Nrf2 nuclear transfer, and regulating the PI3Kα/Akt/mTOR/HIF-1α signaling pathway.
  • Naturally occurring phenolic acid compound
  • Oral effectiveness
  • Anti-inflammatory
  • Antioxidant
  • Anti-cardiac hypertrophy effects
  • Alleviates LPS and ISO by regulating proinflammatory factor expression
  • Inhibits NF-κB activity
  • Promotes Nrf2 nuclear transfer
  • Regulates PI3Kα/Akt/mTOR/HIF-1α signaling pathway
  • Solid appearance, white to off-white color
  • Soluble in DMSO (50 mg/mL)