Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Cryptococcus liquefaciens derived from ATCC™ 34140
SDP

Catalog No. 23045912 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-045-912 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-045-912 Supplier Microbiologics™ Supplier No. 0451K

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Microbiologics Cryptococcus liquefaciens derived from ATCC™ 34140 has a simple, all-inclusive design that makes life easier for lab techs while reducing chances for errors

  • Biosafety Level: 2
  • 6 self-contained units of a single organism
  • Formerly Cryptococcus albidus
  • Each KWIK-STIK contains a qualitative lyophilized microorganism pellet, ampoule of hydrating fluid and inoculating swab

Specifications

ATCC® 34140
Biosafety Level 2
Content And Storage 2°C to 8°C
Format KWIK-STIK
Manufacturing Origin United States
For Use With (Application) Microbial Identification
Packaging Type Plastic Canister
Quantity 6
Species Cryptococcus liquefaciens
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.