Learn More
3B Scientific™ Cryptogamae Elementary Slide Set
SDPSCI_ED
Catalog No. S95045L Shop All 3B Scientific Products
Change view
Click to view available options
Quantity:
25
Catalog No. Quantity
S95045L 25
1 options
Catalog No. S95045L Supplier 3B Scientific™ Supplier No. W13425
Add to Cart
Add to Cart
25 microscope slides
  • 1(e). Bacteria type slide shows cocci, bacilli, spirilli
  • 2(c). Oscillatoria, blue green alga
  • 3(c). Pleurococcus, green alga
  • 4(d). Eudorina, small colonies
  • 5(c). Diatoms, mixed species
  • 6(e). Spirogyra in conjugation with zygotes
  • 7(d). Fucus, brown alga, female conceptacle with oogonia t.s.
  • 8(d). Fucus, male conceptacle with antheridia t.s.
  • 9(c). Mucor, black mold, mycelium and sporangia
  • 10(c). Peziza, apothecium with asci t.s.
  • 11(e). Claviceps purpurea, ergot, stroma with perithecia l.s.
  • 12(c). Morchella, morel, fruiting body t.s.
  • 13(b). Saccharomyces, yeast, budding
  • 14(c). Psalliota, gill fungus, pileus with lamellae t.s.
  • 15(c). Coprinus, mushroom, t.s. typical basidia and spores
  • 16(d). Lobaria pulmonaria, foliose lichen, thallus with symbiotic algae t.s.
  • 17(d). Moss stem with leaves w.m.
  • 18(d). Marchantia, liverwort, thallus with cupule and gemmae l.s.
  • 19(d). Marchantia, antheridia l.s.
  • 20(d). Marchantia, archegonia l.s.
  • 21(d). Polytrichum, moss, capsule with spores t.s.
  • 22(d). Equisetum, horsetail, strobilus with spores l.s.
  • 23(c). Aspidium (Dryopteris), fern, stem t.s.
  • 24(d). Aspidium, leaf with sporangia and spores t.s.
  • 25(d). Fern prothallium w.m.
Specifications
Type Cryptogamae
Quantity 25