Learn More
StatLab Cryptosporidium Control Slides
Description
- Cryptosporidium Control Slides contain high organism counts on positive and negative wells and satisfy regulatory requirements for Cryptosporidium
- Use with Crypto Kinyoun Stain Sets
- Positive well: formalin fixed cryptosporidium
- Negative well: formalin fixed normal fecal flora
- 10 slides/box
Specifications
Specifications
| Product Type | Control Slide |
| Quantity | 10 Slides/Box |