Learn More
ATCC Cryptosporidium parvum; Quantitative DNA
Catalog No. 502382008
Encompass
Quantitative Genomic DNA from Cryptosporidium parvum can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine protozoan load and for other molecular biology applications.
Catalog No. 50-238-2008 Supplier ATCC Supplier No. 502382008
Quantitative Genomic DNA from Cryptosporidium parvum can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine protozoan load and for other molecular biology applications.