Learn More
Selleck Chemical LLC Cryptotanshinone (Tanshinone C) S2285-10mM/1mL
SDP
Catalog No. 503112712
Encompass_Preferred
Cryptotanshinone (Tanshinone C) is a STAT3 inhibitor with IC50 of 4 6 M in a cell-free assay strongly inhibits phosphorylation of STAT3 Tyr705 with a small effect on STAT3 Ser727 but none against STAT1 nor STAT5 Cryptotanshinone induces ROS-dependent autophagy and mitochondria-mediated apoptosis
Catalog No. 50-311-2712 Supplier Selleck Chemical LLC Supplier No. S228510MM/1ML
Cryptotanshinone (Tanshinone C) is a STAT3 inhibitor with IC50 of 4 6 M in a cell-free assay strongly inhibits phosphorylation of STAT3 Tyr705 with a small effect on STAT3 Ser727 but none against STAT1 nor STAT5 Cryptotanshinone induces ROS-dependent autophagy and mitochondria-mediated apoptosis