Learn More
TARGETMOL CHEMICALS INC Cryptotanshinone 50MG
SDP
Catalog No. 502879427
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. Cryptotanshinone (Cryptotanshinon), a STAT3 inhibitor (IC50 4.6 uM) in a cell-free assay, strongly inhibits phosphorylation of STAT3 Tyr705, with a little effect on STAT3 Ser727, but no inhibition for STAT1 nor STAT5. Purity 98.1%
Catalog No. 50-287-9427 Supplier TARGETMOL CHEMICALS INC Supplier No. T281450MG
Also available in 1 mL, 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. Cryptotanshinone (Cryptotanshinon), a STAT3 inhibitor (IC50 4.6 uM) in a cell-free assay, strongly inhibits phosphorylation of STAT3 Tyr705, with a little effect on STAT3 Ser727, but no inhibition for STAT1 nor STAT5. Purity 98.1%